3-[(2,4-Difluorobenzyl)oxy]azetidine

Supplier: Matrix Scientific

049580-500MG 049580-2.5G
101888-224EA 607.69 USD
101888-224 101888-226
3-[(2,4-Difluorobenzyl)oxy]azetidine
3-[(2,4-Difluorobenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₀H₁₁F₂NO
MDL Number: MFCD11874679
CAS Number: 1121592-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR