3-[(4-Bromobenzyl)oxy]azetidine trifluoroacetate

Supplier: Matrix Scientific

049578-500MG 049578-2.5G
101888-216EA 364.62 USD
101888-216 101888-218
3-[(4-Bromobenzyl)oxy]azetidine trifluoroacetate
3-[(4-Bromobenzyl)oxy]azetidine trifluoroacetate

Hazard: Irritant

Formula: C₁₂H₁₃BrF₃NO₃
MDL Number: MFCD22581087
CAS Number: 1425040-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR