Methyl-4-[(3-azetidinyloxy)methyl]benzoate

Supplier: Matrix Scientific

049576-500MG 049576-2.5G
101888-208EA 364.62 USD
101888-208 101888-210
Methyl-4-[(3-azetidinyloxy)methyl]benzoate
Methyl-4-[(3-azetidinyloxy)methyl]benzoate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD11874424
CAS Number: 1121587-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR