Methyl-2-[(3-azetidinyloxy)methyl]benzoate

Supplier: Matrix Scientific

049575-500MG 049575-2.5G
101888-204EA 364.62 USD
101888-204 101888-206
Methyl-2-[(3-azetidinyloxy)methyl]benzoate
Methyl-2-[(3-azetidinyloxy)methyl]benzoate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD13559986
CAS Number: 1220021-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR