3-(4-Bromo-2-methylphenoxy)azetidine

Supplier: Matrix Scientific

049574-500MG 049574-2.5G
101888-200EA 364.62 USD
101888-200 101888-202
3-(4-Bromo-2-methylphenoxy)azetidine
3-(4-Bromo-2-methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD09861886
CAS Number: 954223-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR