3-Azetidinyl-2-methoxy-4-propylphenyl ether

Supplier: Matrix Scientific

049573-500MG 049573-2.5G
101888-196EA 364.62 USD
101888-196 101888-198
3-Azetidinyl-2-methoxy-4-propylphenyl ether
3-Azetidinyl-2-methoxy-4-propylphenyl ether

Hazard: Irritant

Formula: C₁₃H₁₉NO₂
MDL Number: MFCD13559985
CAS Number: 1219982-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR