3-(2-Benzyl-4-chlorophenoxy)azetidine

Supplier: Matrix Scientific

049572-500MG 049572-2.5G
101888-192EA 441.54 USD
101888-192 101888-194
3-(2-Benzyl-4-chlorophenoxy)azetidine
3-(2-Benzyl-4-chlorophenoxy)azetidine

Hazard: Irritant

Formula: C₁₆H₁₆ClNO
MW: 273.77 g/mol
MDL Number: MFCD13559984
CAS Number: 1219963-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR