3-(2,4-Dichloro-3,5-dimethylphenoxy)azetidine

Supplier: Matrix Scientific

049571-500MG 049571-2.5G
101888-188EA 364.62 USD
101888-188 101888-190
3-(2,4-Dichloro-3,5-dimethylphenoxy)azetidine
3-(2,4-Dichloro-3,5-dimethylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₁H₁₃Cl₂NO
MDL Number: MFCD13559983
CAS Number: 1219982-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR