3-(3-Ethoxyphenoxy)azetidine hydrochloride
Supplier: Matrix Scientific
049569-500MG
049569-2.5G
101888-180EA
364.62
USD
101888-180
101888-182
3-(3-Ethoxyphenoxy)azetidine hydrochloride
3-(3-Ethoxyphenoxy)azetidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆ClNO₂ |
MDL Number:
MFCD28291765 CAS Number: 1820684-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...