3-(2-Iodophenoxy)azetidine

Supplier: Matrix Scientific

049568-500MG 049568-2.5G
101888-176EA 364.62 USD
101888-176 101888-178
3-(2-Iodophenoxy)azetidine
3-(2-Iodophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀INO
MDL Number: MFCD13559980
CAS Number: 1219982-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR