3-(4-Bromo-2-fluorophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049567-500MG 049567-2.5G
101888-172EA 364.62 USD
101888-172 101888-174
3-(4-Bromo-2-fluorophenoxy)azetidine hydrochloride
3-(4-Bromo-2-fluorophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀BrClFNO
MDL Number: MFCD28013198
CAS Number: 1713160-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR