3-(2-Nitrophenoxy)azetidine

Supplier: Matrix Scientific

049566-500MG 049566-2.5G
101888-168EA 364.62 USD
101888-168 101888-170
3-(2-Nitrophenoxy)azetidine
3-(2-Nitrophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MDL Number: MFCD11848759
CAS Number: 1219948-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR