3-(4-Isopropyl-3-methylphenoxy)azetidine

Supplier: Matrix Scientific

049565-500MG 049565-2.5G
101888-164EA 364.62 USD
101888-164 101888-166
3-(4-Isopropyl-3-methylphenoxy)azetidine
3-(4-Isopropyl-3-methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD13559978
CAS Number: 1219963-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR