3-(2-Bromo-4-fluorophenoxy)azetidine

Supplier: Matrix Scientific

049564-500MG 049564-2.5G
101888-160EA 364.62 USD
101888-160 101888-162
3-(2-Bromo-4-fluorophenoxy)azetidine
3-(2-Bromo-4-fluorophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₉BrFNO
MDL Number: MFCD09861951
CAS Number: 954225-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR