3-(3,4-Difluorophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049563-500MG 049563-2.5G
101888-156EA 384.62 USD
101888-156 101888-158
3-(3,4-Difluorophenoxy)azetidine hydrochloride
3-(3,4-Difluorophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀ClF₂NO
MDL Number: MFCD21605756
CAS Number: 954220-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR