3-(2,4-Difluorophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049562-500MG 049562-2.5G
101888-152EA 166.15 USD
101888-152 101888-154
3-(2,4-Difluorophenoxy)azetidine hydrochloride
3-(2,4-Difluorophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀ClF₂NO
MDL Number: MFCD16872040
CAS Number: 1241675-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR