3-(3-Methyl-4-nitrophenoxy)azetidine

Supplier: Matrix Scientific

049561-500MG 049561-2.5G
101888-148EA 364.62 USD
101888-148 101888-150
3-(3-Methyl-4-nitrophenoxy)azetidine
3-(3-Methyl-4-nitrophenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD13559977
CAS Number: 1220029-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR