3-(4-Nitrophenoxy)azetidine

Supplier: Matrix Scientific

049560-500MG 049560-2.5G
101888-144EA 364.62 USD
101888-144 101888-146
3-(4-Nitrophenoxy)azetidine
3-(4-Nitrophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀N₂O₃
MDL Number: MFCD11848758
CAS Number: 1219977-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR