3-(4-Methyl-2-nitrophenoxy)azetidine

Supplier: Matrix Scientific

049558-500MG 049558-2.5G
101888-136EA 364.62 USD
101888-136 101888-138
3-(4-Methyl-2-nitrophenoxy)azetidine
3-(4-Methyl-2-nitrophenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD13559976
CAS Number: 1219982-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR