3-Azetidinyl-8-quinolinyl ether dihydrochloride

Supplier: Matrix Scientific

049557-500MG 049557-2.5G
101888-132EA 364.62 USD
101888-132 101888-134
3-Azetidinyl-8-quinolinyl ether dihydrochloride
3-Azetidinyl-8-quinolinyl ether dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄Cl₂N₂O
MDL Number: MFCD24849140
CAS Number: 1449117-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR