3-(3-Azetidinyloxy)pyridine dihydrochloride

Supplier: Matrix Scientific

049556-500MG 049556-2.5G
101888-128EA 364.62 USD
101888-128 101888-130
3-(3-Azetidinyloxy)pyridine dihydrochloride
3-(3-Azetidinyloxy)pyridine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₂Cl₂N₂O
MDL Number: MFCD18089813
CAS Number: 1354543-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR