3-(2-Naphthyloxy)azetidine

Supplier: Matrix Scientific

049555-500MG 049555-2.5G
101888-124EA 453.85 USD
101888-124 101888-126
3-(2-Naphthyloxy)azetidine
3-(2-Naphthyloxy)azetidine

Hazard: Irritant

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
MDL Number: MFCD13559974
CAS Number: 784123-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR