3-(1-Naphthyloxy)azetidine

Supplier: Matrix Scientific

049553-500MG 049553-2.5G
101888-116EA 364.62 USD
101888-116 101888-118
3-(1-Naphthyloxy)azetidine
3-(1-Naphthyloxy)azetidine

Hazard: Irritant

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
MDL Number: MFCD13559972
CAS Number: 782433-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR