3-Azetidinyl-2,3-dihydro-1H-inden-5-yl ether hydrochloride
Supplier: Matrix Scientific
049552-500MG
049552-2.5G
101888-112EA
364.62
USD
101888-112
101888-114
3-Azetidinyl-2,3-dihydro-1H-inden-5-yl ether hydrochloride
3-Azetidinyl-2,3-dihydro-1H-inden-5-yl ether hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₆ClNO |
MDL Number:
MFCD25970561 CAS Number: 1449117-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...