3-Azetidinyl-2-methoxy-4-methylphenyl ether

Supplier: Matrix Scientific

049548-500MG 049548-2.5G
101888-096EA 364.62 USD
101888-096 101888-098
3-Azetidinyl-2-methoxy-4-methylphenyl ether
3-Azetidinyl-2-methoxy-4-methylphenyl ether

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD13559967
CAS Number: 1219948-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR