3-(4-Methylphenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049547-500MG 049547-2.5G
101888-092EA 170.77 USD
101888-092 101888-094
3-(4-Methylphenoxy)azetidine hydrochloride
3-(4-Methylphenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MW: 199.68 g/mol
MDL Number: MFCD16613952
CAS Number: 1228070-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR