3-[4-(sec-Butyl)phenoxy]azetidine

Supplier: Matrix Scientific

049546-500MG 049546-2.5G
101888-088EA 364.62 USD
101888-088 101888-090
3-[4-(sec-Butyl)phenoxy]azetidine
3-[4-(sec-Butyl)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD13559966
CAS Number: 1219977-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR