3-(4-Isopropylphenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049545-500MG 049545-2.5G
101888-084EA 364.62 USD
101888-084 101888-086
3-(4-Isopropylphenoxy)azetidine hydrochloride
3-(4-Isopropylphenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD24369563
CAS Number: 1452484-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR