3-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]azetidine
Supplier: Matrix Scientific
049543-500MG
049543-2.5G
101888-076EA
364.62
USD
101888-076
101888-078
3-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]azetidine
3-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]azetidine
Hazard: Irritant
Formula:
C₁₇H₂₇NO |
MDL Number:
MFCD13559963 CAS Number: 1219982-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...