3-[4-(tert-Butyl)phenoxy]azetidine hydrochloride

Supplier: Matrix Scientific

049542-500MG 049542-2.5G
101888-072EA 338.46 USD
101888-072 101888-074
3-[4-(tert-Butyl)phenoxy]azetidine hydrochloride
3-[4-(tert-Butyl)phenoxy]azetidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD24849139
CAS Number: 1449117-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR