3-[4-(Trifluoromethyl)phenoxy]azetidine hydrochloride
Supplier: Matrix Scientific
049540-500MG
049540-2.5G
101888-064EA
364.62
USD
101888-064
101888-066
3-[4-(Trifluoromethyl)phenoxy]azetidine hydrochloride
3-[4-(Trifluoromethyl)phenoxy]azetidine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₀F₃NO·ClH MW: 253.649 g/mol |
MDL Number:
MFCD18071794 CAS Number: 1236862-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...