Methyl-4-(3-azetidinyloxy)benzoate

Supplier: Matrix Scientific

049539-500MG 049539-2.5G
101888-060EA 364.62 USD
101888-060 101888-062
Methyl-4-(3-azetidinyloxy)benzoate
Methyl-4-(3-azetidinyloxy)benzoate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD13559960
CAS Number: 1219948-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR