3-([1,1'-Biphenyl]-4-yloxy)azetidine hydrochloride

Supplier: Matrix Scientific

049538-500MG 049538-2.5G
101888-056EA 364.62 USD
101888-056 101888-058
3-([1,1'-Biphenyl]-4-yloxy)azetidine hydrochloride
3-([1,1'-Biphenyl]-4-yloxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₆ClNO
MDL Number: MFCD24369562
CAS Number: 1186234-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR