3-[4-(Benzyloxy)phenoxy]azetidine

Supplier: Matrix Scientific

049535-500MG 049535-2.5G
101888-044EA 364.62 USD
101888-044 101888-046
3-[4-(Benzyloxy)phenoxy]azetidine
3-[4-(Benzyloxy)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₆H₁₇NO₂
MDL Number: MFCD13559957
CAS Number: 1219977-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR