3-(4-Methoxyphenoxy)azetidine

Supplier: Matrix Scientific

049534-500MG 049534-2.5G
101888-040EA 615.38 USD
101888-040 101888-042
3-(4-Methoxyphenoxy)azetidine
3-(4-Methoxyphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD09756536
CAS Number: 954220-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR