N-[4-(3-Azetidinyloxy)phenyl]acetamide

Supplier: Matrix Scientific

049533-500MG 049533-2.5G
101888-036EA 364.62 USD
101888-036 101888-038
N-[4-(3-Azetidinyloxy)phenyl]acetamide
N-[4-(3-Azetidinyloxy)phenyl]acetamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MDL Number: MFCD13559956
CAS Number: 1219961-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR