3-(4-Iodophenoxy)azetidine

Supplier: Matrix Scientific

049532-500MG 049532-2.5G
101888-032EA 364.62 USD
101888-032 101888-034
3-(4-Iodophenoxy)azetidine
3-(4-Iodophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀INO
MDL Number: MFCD13559955
CAS Number: 1219948-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR