3-(4-Chloro-3,5-dimethylphenoxy)azetidine

Supplier: Matrix Scientific

049531-500MG 049531-2.5G
101888-028EA 364.62 USD
101888-028 101888-030
3-(4-Chloro-3,5-dimethylphenoxy)azetidine
3-(4-Chloro-3,5-dimethylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₁H₁₄ClNO
MW: 211.69 g/mol
MDL Number: MFCD13559954
CAS Number: 1219977-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR