3-(4-Chloro-3-methylphenoxy)azetidine

Supplier: Matrix Scientific

049530-500MG 049530-2.5G
101888-024EA 364.62 USD
101888-024 101888-026
3-(4-Chloro-3-methylphenoxy)azetidine
3-(4-Chloro-3-methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MDL Number: MFCD13559953
CAS Number: 1219948-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR