3-(2-Bromo-4-chlorophenoxy)azetidine

Supplier: Matrix Scientific

049529-500MG 049529-2.5G
101888-020EA 364.62 USD
101888-020 101888-022
3-(2-Bromo-4-chlorophenoxy)azetidine
3-(2-Bromo-4-chlorophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₉BrClNO
MW: 262.54 g/mol
MDL Number: MFCD09861959
CAS Number: 954225-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR