3-(4-Chlorophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049528-500MG 049528-2.5G
101888-016EA 692.31 USD
101888-016 101888-018
3-(4-Chlorophenoxy)azetidine hydrochloride
3-(4-Chlorophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₀ClNO·ClH
MW: 220.096 g/mol
MDL Number: MFCD00277797
CAS Number: 490021-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR