3-(4-Fluorophenoxy)azetidine

Supplier: Matrix Scientific

049527-500MG 049527-2.5G
101888-012EA 364.62 USD
101888-012 101888-014
3-(4-Fluorophenoxy)azetidine
3-(4-Fluorophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₁₀FNO
MDL Number: MFCD09861914
CAS Number: 702628-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR