3-(4-Bromophenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049526-500MG 049526-2.5G
101888-008EA 692.31 USD
101888-008 101888-010
3-(4-Bromophenoxy)azetidine hydrochloride
3-(4-Bromophenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClNO
MDL Number: MFCD05664840
CAS Number: 1380571-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR