3-(2-Isopropyl-5-methylphenoxy)azetidine
Supplier: Matrix Scientific
049524-500MG
049524-2.5G
101888-000EA
364.62
USD
101888-000
101888-002
3-(2-Isopropyl-5-methylphenoxy)azetidine
3-(2-Isopropyl-5-methylphenoxy)azetidine
Hazard: Irritant
Formula:
C₁₃H₁₉NO |
MDL Number:
MFCD13559952 CAS Number: 1220028-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...