3-(3,5-Dimethylphenoxy)azetidine

Supplier: Matrix Scientific

049523-500MG 049523-2.5G
101887-996EA 364.62 USD
101887-996 101887-998
3-(3,5-Dimethylphenoxy)azetidine
3-(3,5-Dimethylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD13559951
CAS Number: 1219961-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR