3-(3,4-Dimethylphenoxy)azetidine hydrochloride

Supplier: Matrix Scientific

049522-500MG 049522-2.5G
101887-992EA 364.62 USD
101887-992 101887-994
3-(3,4-Dimethylphenoxy)azetidine hydrochloride
3-(3,4-Dimethylphenoxy)azetidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD24849138
CAS Number: 1449117-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR