3-(3-Methylphenoxy)azetidine

Supplier: Matrix Scientific

Synonyms: 3-(3-Methylphenoxy)azetidine hydrochloride

049521-500MG 049521-2.5G
101887-988EA 364.62 USD
101887-988 101887-990
3-(3-Methylphenoxy)azetidine
3-(3-Methylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₀H₁₃NO
MDL Number: MFCD09861892
CAS Number: 949100-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR