3-(3-Isopropylphenoxy)azetidine

Supplier: Matrix Scientific

049520-500MG 049520-2.5G
101887-984EA 364.62 USD
101887-984 101887-986
3-(3-Isopropylphenoxy)azetidine
3-(3-Isopropylphenoxy)azetidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD13559949
CAS Number: 1219982-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR