3-[3-(Trifluoromethyl)phenoxy]azetidine hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-[3-(Trifluoromethyl)phenoxy]-azetidine hydrochloride

049518-500MG 049518-2.5G
101887-976EA 364.62 USD
101887-976 101887-978
3-[3-(Trifluoromethyl)phenoxy]azetidine hydrochloride
3-[3-(Trifluoromethyl)phenoxy]azetidine hydrochloride

Hazard: Irritant

Formula: C10H11ClF3NO
MW: 253.65 g/mol
MDL Number: MFCD05664837
CAS Number: 1188374-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR