Methyl-3-(3-azetidinyloxy)benzoate

Supplier: Matrix Scientific

049516-500MG 049516-2.5G
101887-968EA 364.62 USD
101887-968 101887-970
Methyl-3-(3-azetidinyloxy)benzoate
Methyl-3-(3-azetidinyloxy)benzoate

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD13559947
CAS Number: 1219976-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR